Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SKF 77434 hydrobromide, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective dopamine D1-like receptor partial agonist (IC50 values are 19.7 and 2425 nM for binding to D1-like and D2-like receptors respectively). Centrally active following systemic administration in vivo.
Specifications
Quantity | 50 mg |
Formula Weight | 376.29 |
Percent Purity | >99% |
Chemical Name or Material | SKF 77434 hydrobromide |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction