Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynamic Diagnostics Sink Splash Guard with Easy Cut-Away
Bio hazard safety shield
$216.00 - $239.00
Specifications
Dimensions (L x W x H) | 18.75 x 4.5 x 18 in. |
---|---|
Height (English) | 18 in. |
Length (English) | 18.75 in. |
Size | Small |
Material | Acrylic |
Catalog Number | Mfr. No. | Length (English) | Dimensions (L x W x H) | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Dimensions (L x W x H) | Size | Price | Quantity & Availability | |||
22-899-672
|
Dynamic Diagnostics
4000148 |
18.75 in. | 18.75 x 4.5 x 18 in. | Small |
Each for $216.00
|
|
|||
22-899-673
|
Dynamic Diagnostics
4000149 |
23.625 in. | 23.625 x 4.5 x 18 in. | Medium |
Each for $239.00
|
|
|||
Description
- Easy cut-away
Specifications
18.75 x 4.5 x 18 in. | |
18.75 in. | |
Acrylic |
18 in. | |
Small |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title