Learn More
Pendotech Pendotech Single Use Turbidity Sensor
Supplier: Pendotech SPECPS8806CM
Single Use Turbidity Sensor, 6.5cm path length, non-sterile, polysulfone, 3/4 inch Sanitary Flange Inlet/Outlet