Learn More
Pendotech Pendotech 3/4 Inch Sanitary Flange To 3/8 Inch Hose Barb Single Use Pressure Sensor
Supplier: Pendotech PREPSN5038
Single Use Pressure Sensor, non-sterile, polysulfone, 3/4 inch Sanitary Flange to 3/8” hose barb