Learn More
Pendotech Pendotech 1/2 Inch Hose Barb Single Use Rotary Flow Meter
Supplier: Pendotech FM23WV
Single Use Rotary Flow Meter, non-sterile, PVDF, 1/2 inch hose barb, 1.0-20.0 lpm, clip mount. With individual calibrations.