Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Simulaids™ Rescue Randy Combat Challenge and I.A.F.F. Manikins
Official manikin of the Firefighter Combat Challenge
Supplier: Simulaids™ 1491475
This item is not returnable.
View return policy
Description
Developed for lifelike adult or juvenile victim handling, transportation, and extrication training.
- Used by the U.S. military, fire, and police departments, safety teams, and emergency personnel for rescue and extrication from pole top, confined spaces, collapsed buildings, smoke rooms and ladder carry-down protocols worldwide
- Can be safely used in situations too hazardous or uncomfortable for human volunteers
- Made of durable vinyl with 4100 lb. test plastic-coated cables
- Features include articulated joints and weight distribution according to human weight distribution chart
- Size: 56 × 27 × 13 in.
Specifications
I.A.F.F. Rescue Randy Manikin | |
Developed for Lifelike Adult or Juvenile Victim Handling, Transportation and Extrication Training | |
Articulated joints and weight distribution according to human weight distribution chart | |
Vinyl, Steel |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction