Learn More
Structure Probe Inc SPI Silver Streaker for SEM applications, 30g
Supplier: Structure Probe Inc 05005AB
The SPI Supplies Silver Streaker is a unique silver paint application system, ideal for mounting powders, granules, particles, and anything requiring minimum exposure to liquids or vapors. Pressure is applied to the sponge top applicator which then releases a thin, even discharge of silver paint. This layer is extremely thin, evaporates literally in a flash, and uses minimal silver solids for each application of the paint. The SPI Silver Streaker was actually an invention of the early workers in the Structure Probe SEM analytical services laboratories where the need for this kind of product was readily recognized.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.