Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver nitrate, TraceSELECT™, ≥99.998% (metals basis), Honeywell™ Flula™

Catalog Number 60046856
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O

TraceSELECT™, for trace analysis,≥99.998% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY
Quantity 25 g
Linear Formula AgNO3
UN Number UN1493
Formula Weight 169.87g/mol
Percent Purity ≥99.998% (metals basis)
Grade TraceSELECT™
Packaging Glass Bottle
Density Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver nitrate
Hazard Statement P210-P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation 5.1/PGII
RTECSNumber 231-853-9
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit