Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaSep™ Flash Cartridges, Silica-Based, Tosic Acid (SCX), 40 to 63 μm, 60 Å

SiliaSep™ Flash Cartridges, Silica-Based, Tosic Acid (SCX), 40 - 63 μm, 60 Å (4 g to 1600 g) are packed with Tosic Acid sorbent (SCX) for purification of weak basic molecules.

$106.00 - $8365.00

Specifications

For Use With (Application) Purification of weak basic molecules
Material Polypropylene
Type Flash Cartridge
Carbon Load 9%
Content And Storage Keep dry.
View More Specs

Products 16
Catalog Number Mfr. No. Column Type Diameter (Metric) Outer Max. Pressure Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Column Type Diameter (Metric) Outer Max. Pressure Quantity Price Quantity & Availability  
01-183-802
View Documents
SiliCycle™
FLHR60530BISO04
4 g Flash Cartridge 12 mm 225 psi/16 bar 2 Pk.
Pack of 2 for $106.00
Add to Cart
 
01-183-803
View Documents
SiliCycle™
FLHR60530BISO04B
4 g Flash Cartridge 12 mm 225 psi/16 bar 10 Pk.
Pack of 10 for $446.00
Add to Cart
 
01-183-804
View Documents
SiliCycle™
FLHR60530BISO12
12 g Flash Cartridge 21 mm 225 psi/16 bar 1 Pk.
Pack for $153.00
Add to Cart
 
01-183-805
View Documents
SiliCycle™
FLHR60530BISO12B
12 g Flash Cartridge 21 mm 225 psi/16 bar 10 Pk.
Pack of 10 for $1,246.00
Add to Cart
 
01-183-806
View Documents
SiliCycle™
FLHR60530BISO25
25 g Flash Cartridge 21 mm 225 psi/16 bar 1 Pk.
Pack for $231.00
Add to Cart
 
01-183-807
View Documents
SiliCycle™
FLHR60530BISO25B
25 g Flash Cartridge 21 mm 225 psi/16 bar 10 Pk.
Pack of 10 for $1,957.00
Add to Cart
 
01-183-808
View Documents
SiliCycle™
FLHR60530BISO40
40 g Flash Cartridge 27 mm 225 psi/16 bar 1 Pk.
Pack for $274.00
Add to Cart
 
01-183-809
View Documents
SiliCycle™
FLHR60530BISO40B
40 g Flash Cartridge 27 mm 225 psi/16 bar 5 Pk.
Pack of 5 for $1,157.00
Add to Cart
 
01-183-810
View Documents
SiliCycle™
FLHR60530BISO80
80 g Flash Cartridge 31 mm 225 psi/16 bar 1 Pk.
Pack for $493.00
Add to Cart
 
01-183-811
View Documents
SiliCycle™
FLHR60530BISO80B
80 g Flash Cartridge 31 mm 225 psi/16 bar 5 Pk.
Pack of 5 for $2,094.00
Add to Cart
 
01-183-797
View Documents
SiliCycle™
FLHR60530BIS120
120 g Flash Cartridge 36 mm 205 psi/13 bar 1 Pk.
Pack for $691.00
Add to Cart
 
01-183-798
View Documents
SiliCycle™
FLHR60530BIS120B
120 g Flash Cartridge 36 mm 205 psi/13 bar 5 Pk.
Pack of 5 for $2,944.00
Add to Cart
 
01-183-799
View Documents
SiliCycle™
FLHR60530BIS220
220 g Flash Cartridge 60 mm 160 psi/11 bar 1 Pk.
Pack for $1,291.00
Add to Cart
 
01-183-800
View Documents
SiliCycle™
FLHR60530BIS330
330 g Flash Cartridge 60 mm 160 psi/11 bar 1 Pk.
Pack for $1,947.00
Add to Cart
 
01-183-801
View Documents
SiliCycle™
FLHR60530BIS750
800 g Flash Cartridge 78 mm 125 psi/8 bar 1 Pk.
Pack for $4,183.00
Add to Cart
 
01-183-796
View Documents
SiliCycle™
FLHR60530BI1500
1600 g Flash Cartridge 104 mm 100 psi/7 bar 1 Pk.
Pack for $8,365.00
Add to Cart
 
Description

Description

  • Directly compatible with a variety of flash systems (slip tip connection type, but can be used with many luer lock systems)
  • Pre-packed disposable flash cartridges
  • Unique one-piece polypropylene cartridge for high pressure and solvent resistance
  • Leak-free guaranteed
  • Made with high quality silica (40 - 63 μm, 60 Å)
  • Tight particle size distribution
  • Low fines
  • Neutral pH
  • Controlled water content
  • Very diversified formats (4, 12, 25, 40, 80, 120, 220, 330, 800, and 1600 g)
  • Effective packing technology
  • Good resolution, no tailing
  • Stringent quality control (ISO 9001:2015 Registered)
  • Lot-to-lot reproducibility
  • For 800 g and 1600 g XL formats, an adapter may be needed to connect the cartridge to the system
Specifications

Specifications

Purification of weak basic molecules
Flash Cartridge
Keep dry.
Yes
2 to 10
SiliaSep™
500 m2/g
Polypropylene
9%
40 to 63 μm
Irregular
60 Å
Tosic Acid (Ethyltosic Acid, SCX)
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit