Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Diagnostics™ Sigmacote™
Reacts with surface silanol groups on glass to produce a neutral, hydrophobic, microscopically thin film
$83.01 - $155.52
Specifications
Color | Colorless |
---|---|
Content And Storage | 2°C to 8°C |
Type | Siliconizing Reagent for Glass and Other Surfaces |
Intended Use | Used to treat GC injection glass inserts. |
Form | Solution |
Description
- Clear, colorless solution of a chlorinated organopolysiloxane in heptane
Specifications
Colorless | |
Siliconizing Reagent for Glass and Other Surfaces | |
Solution |
2°C to 8°C | |
Used to treat GC injection glass inserts. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title