Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Diagnostics™ Poly-L-lysine Solution
Can be used in promoting cell adhesion to solid substrates
Supplier: Sigma Diagnostics™ P8920100ML
Includes: Can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations
Description
Components- Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue
- The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water
- A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties
Specifications
25988-63-0 | |
0.1% (w/v) in H2O | |
Can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations | |
100mL |
Colorless | |
Solution | |
Poly-L-lysine solution | |
2–8°C |
Safety and Handling
ShelfLife : Expiry date on the label.
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction