Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Diagnostics™ Phosphomolybdic Acid Solution
For use in Masson Trichrome procedure
Supplier: Sigma Diagnostics™ HT153250ML
Description
- Concentration: 10%
- Formation of precipitate does not affect performance
Specifications
12026-57-2 | |
Solution | |
Sigma Masson trichrome procedure | |
Phosphomolybdic acid solution | |
Molybdophosphoric acid |
Faint Yellow to Dark Yellow | |
1,825.25g/mol | |
H3Mo12O40P | |
250mL |
Safety and Handling
ShelfLife : Expiry date on the label
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro diagnostic use
Spot an opportunity for improvement?
Provide Content Correction