Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Sigma-Aldrich™ Flash Silica (Dried, Spherical)
High-purity grade
$113.00 - $655.00
Specifications
For Use With (Application) | Flash Chromatography |
---|---|
Product Line | Sigma-Aldrich |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
11-101-0960
|
MilliporeSigma™ Supelco™
97728U |
100 g |
Each for $113.00
|
|
|||||
11-100-8788
|
MilliporeSigma™ Supelco™
97729U |
1 kg |
Each for $655.00
|
|
|||||
Description
- Pore Size: 0.7-0.9 cm3/g pore volume
- Surface Area: 480 m2/g
- MDL Number: MFCD00011232
- BP: 2230°C
- MP: gt;1600°C
- CAS: 112926-00-8
Specifications
Flash Chromatography | |
Sigma-Aldrich |
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title