Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthineers Wright-Giemsa Stain Pack for HEMA-TEK™ 3000 Slide Stainer
For use with HEMA-TEK™ 3000 Slide Stainer.
Supplier: Siemens Healthineers 10314755
Description
- Contains 1 bottle each of stain, buffer and rinse solution
Specifications
Wright-Giemsa Stain Pack | |
Hema-Tek™ 3000 slide stainer | |
1 bottle each of stain, buffer and rinse solution |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction