Learn More
General Data Healthcare™ SHUR/Mount™ Liquid Cover Glass and Mounting Medium
Convenient twist-top spout eliminates messy bottles and droppers
Supplier: General Data Healthcare™ LCA
Description
- Antioxidant prevents fading and UV absorbent prevents yellowing
- Toluene-based acrylic resin allows rapid drying for hand coverslipping
- Xylene-based for automated coverslipping instruments
- Low viscosity minimizes bubble formation under cover glass
Specifications
Automated Coverslippers | |
16 oz. | |
Low Viscosity Mounting Medium | |
471 mL |