Learn More
Sheldon Manufacturing Inc SHELLAB SHAKING WATER BATH 27L
SDP
Supplier:  Sheldon Manufacturing Inc SWBR27
Encompass_Preferred
WATER BATH, RECIPROCATING/SHAKING, 27 Liter, with cover, 115V. Digitally-controlled reciprocating waterbath. Temperature range from Ambient +5°C to 80°C, Oscillation Rate 20-200 RPM. SHEL LAB Reciprocating Water Baths deliver precise temperature control and a smooth reciprocal shaking motion. Temperature and oscillation controls feature an easy to read P.I.D. microprocessor temperature controller and an independent safety thermostat. The bath may be used with or without oscillation. Shaking speed is variable from 20 to 200 oscillations per minute, and the stroke length can be adjusted to three separate settings: 0.5, 1.0, or 1.5 inches to minimize the chance of accidental spillage. Also, the unique basket support system is designed to eliminate tank wear, yet still provide full support and free action for a heavily loaded basket. More information available at www.Shellab.com
Catalog No. NC0812996