Learn More
Colorado Serum Co SHEEP BLOOD DEFIBRONATED 100ML
Supplier: Colorado Serum Co 31123
SHEEP BLOOD DEFIBRONATED,100ml,Defibrinated: Whole blood which has been processed to remove fibrin.There are no additives or preservatives in this product,All blood products are tested for sterility using BHI,Thioglycollate and Tryptic Soy