Learn More
QUAD FIVE SHEEP BLOOD 1OOML DEFIBRINATED
Supplier:  QUAD FIVE 610100
Encompass
DONOR STERILE DEFIBRINATED WHOLE BLOOD, 100 ml, Hematocrit: > 35%, Osmotic Fragility: 0% hemolysis in 0.65% saline solution
Catalog No. NC9640902