Learn More
Epredia™ Shandon™ Uni-Capsettes
Hold tissue specimens during processing and embedding
$257.00
Specifications
Pore Style | Slots |
---|---|
Closure Type | Separate Lid |
Product Type | Processing Cassette |
Description
- Pore style: small slots
- Supplied in bags of 500 cassettes with lids packaged separately
- Available in 10 colors
Specifications
Slots | |
Separate Lid | |
Processing Cassette |