Learn More
Epredia™ Embedding Tissue Cassettes
Hold and identify tissue samples in processing, embedding, and sectioning procedures with Epredia™ Embedding Tissue Cassettes.
Supplier: Epredia™ B1003500LI
Description
- Designed to accept metal lids
- Designed with round hole pores
- Supplied in boxes of 500 cassettes
Specifications
Round Holes | |
Lilac | |
Tissue Cassette |