Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Accell PRDM1 siRNA Reagents - Human, Set of 4, 20nmol
Supplier: HORIZON DISCOVERY BIOSCIENCES LIMITED EQ009322000020
Accell siRNA achieves what no other RNAi product can claim: delivery into difficult-to-transfect cells without transfection reagents, virus, or instrumentation. Researchers around the world are achieving targeted gene silencing in neuronal, immune, and primary cells that had previously been beyond the reach of conventional RNAi products due to toxicity caused by transfection reagents or undesirable viral responses.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.