Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED SET OF 3 SMARTVECTOR INDUCIBLE
Supplier:  HORIZON DISCOVERY BIOSCIENCES LIMITED V3SM767710EG110157
Encompass
Set of 3 SMARTvector Inducible Mouse Raf1 mEF1a-TurboRFP shRNA
Catalog No. NC1745863