Learn More
Binder™ Series VD Vacuum Drying Chambers
SDP
Binder™ Series VD Vacuum Drying Chambers gently dry materials with homogeneous temperature distribution.
Supplier:  Binder™ 90300036
 View more versions of this product
Catalog No. 08-111-023
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • The patented expansion shelf technology ensures optimal heat transfer
  • The shelves are easy to position where required and the inner chamber of the vacuum drying oven is easy to clean
  • APT.line™ preheating chamber technology
  • Controller with time-segment and real-time programming
  • 2 aluminum expansion racks
  • Inert gas connection
  • Shatterproof, spring-mounted safety glass panel
  • Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
  • Computer interface: RS 422
  • 2 relay outputs, 24 V DC (max. 0.4 A)
Specifications
Specifications
15°C to 200°C
44 lb.
2 supplied/5 max.
120 V 60 Hz
Vacuum Drying Chamber
VD 53
13.4 x 15.76 x 15.76 in.
SDS
Documents
Documents
Product Certifications
Accessories