Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS™ Septihol™ Ethyl WFI Sterile Alcohol Solution
A solution of 70% (v/v) reagent-grade alcohol and 30% (v/v) USP WFI designed to remove organic and inorganic residues in aseptic processing areas; double bagged for easy entry into critical environments.
$598.50 - $1236.90
Specifications
Product Type | Ethyl Alcohol |
---|---|
Sterility | Sterile |
Composition | 70% (v/v) Reagent-Grade Alcohol, and 30% (v/v) USP Water for Injection (WFI) |
Filtered | Filtered |
For Use With (Application) | Removing Organic and Inorganic Residues in Aseptic Processing Areas |
Catalog Number | Mfr. No. | Volume (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Price | Quantity & Availability | |||||
19-415-118
|
STERIS™
1S1177 |
22 oz. |
Case of 12 for $882.00
|
|
|||||
19-415-119
|
STERIS™
1S1196 |
32 oz. |
Case of 12 for $1,236.90
|
|
|||||
19-415-120
|
STERIS™
1S1108 |
1 gal. |
Case of 4 for $598.50
|
|
|||||
Description
- Designed to remove organic and inorganic residue from product, and non-product contact surfaces
- A mixture of ethanol, isopropanol, methanol and United States Pharmacopeia (USP) Water For Injection (WFI)
- Free of stabilizers and non-volatile denaturants for quick and complete evaporation
- Ready to use, sterile alcohol eliminates in-house production validation and documentation concerns
- Packages are double bagged for easy entry into controlled environments and aseptic core areas
- Includes extensive documentation package to meet validation objectives
- Each lot documented by a Certificate of Analysis including irradiation and sterility results
- Supported by the STERIS Technical Services team for individualized application and validation assistance
Specifications
Ethyl Alcohol | |
70% (v/v) Reagent-Grade Alcohol, and 30% (v/v) USP Water for Injection (WFI) | |
Removing Organic and Inorganic Residues in Aseptic Processing Areas |
Sterile | |
Filtered |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title