Learn More
Sensorex SENSOREX ORP CALIBERATION KIT
Supplier: Sensorex B125
ORP Calibration Kit,Each ORP Kit contains material for 30 two point calibrations,CONTENTS: 1-pint,pH 4 Buffer; 1-pint pH 7 Buffer;3-4 oz beakers; 75-wood applicators; 1-20 gram bottle quinhydrone;1-instruction sheet