Learn More
Thermo Scientific™ Sensititre™ Gram Negative Urine Plate
Reduce cost per test while achieving accurate, reliable susceptibility results with the three-isolate Thermo Scientific™ Sensititre™ Gram Negative Urine Plate.
Supplier: Thermo Scientific™ YGNUR2F
Description
Reduce cost per test while achieving accurate, reliable susceptibility results with the three-isolate Thermo Scientific™ Sensititre™ Gram Negative Urine Plate.
Reduce cost per test while achieving accurate, reliable susceptibility results with the three-isolate Sensititre Gram Negative Urine Plate.- Comprehensive testing with FDA clearances includes Pseudomonas, Burkholderia, Acinetobacter and Stenotrophomonas spp.
- Dilution ranges meet FDA and CLSI recommendations for non-fermenters, fermenters and Enterobacteriaceae, including expanded coverage of cephalosporins, aztreonam and carbapenems, to cover all in-house validation testing
- Multi-isolate format decreases cost per test
- Automated, semiautomated or manual read options are available for ultimate flexibility
- Extended shelf life and room-temperature storage simplify inventory control
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
H2O (Part No. T3339) | |
MIC | |
Sensititre Gram Negative Urine Plate | |
Gram Negative Urine Plate | |
MIC | |
18 hr | |
10 | |
Autoread or manual | |
50 μL |
Various | |
MHB (Part No. T3462) | |
Various | |
Gram-Negative Organisms | |
35°C O2 | |
3 | |
10/Pk. | |
For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL |