Learn More
Sensidyne™ Detector Tubes: Inorganic Gas Qualitative
Supplier: Sensidyne™ 131
Description
Detector tubes are sold in packs of 10
Specifications
Inorganic Gas Qualitative | |
10 | |
32°F to 104°F |
N/A | |
20 sec. | |
0°C to 40°C |