Learn More
Sensidyne™ Detector Tubes: Acrolein
Detector tubes
Supplier: Sensidyne™ 136
Description
All detector tubes are sold in packs of 10.Specifications
Acrolein | |
1 | |
1 min. | |
0°C to 40°C |
0.005 1.8% | |
10 Standard Tubes | |
32°F to 104°F |