Learn More
Sensidyne™ Compressed Breathing Air (CBA) Analysis Kit
Allows quantitative measurement of four of the common contaminants in breathing air, plus oxygen deficiency.
Supplier: Sensidyne™ 7013911
Description
- Provides quantitative measurement
- Detects oxygen deficiency.
- Kit operates at pressures up to 3000 PSI
- Does not require user calibration
Specifications
CBA Retrofit Kit | |
7013911 | |
1 Ea. |
Quantitatively measure all four common contaminants in breathing air, plus oxygen deficiency | |
3000 psi |