Learn More
PIP™ Semi-Rimless Bifocal Safety Readers
Click to view available options
Magnification Power:
+1.00
+1.50
+2.00
+2.50
+3.00
Description
- 30 mm wide curved top reading magnifier provides the worker with enough lens area to view work throughout the day
- Durable nylon frame with extra wide latex-free rubber nose pads for more comfort when working up close
- Temples with soft rubber pads provide a secure fit
- 8 Base curve lens
- Meets ANSI Z87.1+
Specifications
Description | Extra wide bridge |
Lens Tint | Clear |
Frame Color | Black |
Magnification Power | +1.00 |
Lens Material | Polycarbonate |
Frame Material | Nylon |
UV Protection | 0.9999 |
Warning - Prop 65 - Reproductive Harm