Learn More
Strategic Applications Inc 6"" Trocar for Large Animal Surgery - Semi-Blunt Tip, 7 gauge
SDP
Supplier:  Strategic Applications Inc TRO76
Encompass
Subcutaneous tunneling needles (trocars) offer a quick and easy method for routing catheters and electrodes under the skin. SAI's TRO-7-6 is manufactured from thick-walled medical-grade stainless steel, has a Semi-Blunt 45 beveled tip and is ideal for large animal surgery. Customized tips and lengths are available, please contact us for more information. 7G, 6". Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply.  Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
Catalog No. NC0958974
Due to product restrictions, please to purchase or view availability for this product.