Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ SecureFit™ 3700 Series Protective Eyewear
Click to view available options
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor, Gray
Temple Color:
Blue
Gray
Green
Lime Green
Orange
Description:
Alternate Fit
Alternate Fit with Browguard
Browguard
Regular Fit
Description
- The adjustable ratchet temples provides angle adjustment points on the temple to help create a secure and personalized fit
- Pressure Diffusion Temple (PDT) Technology helps diffuse pressure over the ear for a secure and comfortable fit
- Soft touch nosepiece to help provide all-day comfort
- Polycarbonate Lenses absorb 99.9% of UVA and UVB rays
- Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
- 3M Catalog# SF3701, SF3702, SF3703, SF3707
Specifications
Description | Alternate Fit |
Lens Tint | Clear |
Product Type | Protective eyewear |
For Use With (Application) | Automotive, Machining, Transportation, Sawing, Riveting |
Frame Color | Clear |
No. per Case | 10/Cs. |
Model | SF3701ASGAF-BLU |
Size | Universal |
Temple Style | 3M™ Pressure Diffusion Temple Technology |
Temple Color | Blue |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction