Learn More
VANTE INC SEBRA 1105 Hand Held Sealing Head
Supplier: VANTE INC 1105
MODEL 1105 SEALING HEAD - Handles heavy duty and high capacity demands. The 1105 is compatible with the 2380 – Battery Powered RF Generator or the 2600 – Bench-Top RF Generato. Produces wide tear seals - Easy to clean - Adjusts to changing requirements with no manual intervention - Provides reliability and security for processing of temperature sensitive samples
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.