Learn More
Chemglass Life Sciences Glassblowers Fritted Tube, Straight, Frit 10mm Diameter, Fine Porosity, Tube O.D./13mm
SDP
Supplier:  Chemglass Life Sciences CG20201
Encompass_Preferred
Glassblowers Fritted Tube, Straight, Frit 10mm Diameter, Fine Porosity, Tube O.D./13mm. Cylindrical tube with a centered, sealed in fritted disc, used to construct special apparatus.
Catalog No. CG20201