Learn More
MilliporeSigma SDS AGAR STRIPS 50/PK
Supplier: MilliporeSigma 1442430050
Sabouraud Dextrose Agar Strip; For yeasts and molds found in cleanroom environments, growth is not restricted; Strip Media: SDX; 50/BX