Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SD 2590 hydrochloride
Potent MMP inhibitor
$332.92 - $1394.32
Specifications
Purity | >98% |
---|---|
Description | SD 2590 hydrochloride |
Molecular Weight (g/mol) | 554.96 |
Product Type | SD 2590 hydrochloride |
Description
Potent MMP inhibitor (IC50 values are <0.1, <0.1, 0.18, 1.7, 13 and 28.7 nM for MMP-2, -13, -9, -8, -14 and -3 respectively). Exhibits 100,000-fold selectivity for MMP-2 and -13 over MMP-1. Reduces left ventricular dilation in a myocardial infarction rat model. Orally bioavailable.Specifications
>98% | |
SD 2590 hydrochloride | |
554.96 | |
SD 2590 hydrochloride |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title