Learn More
Enzo Life Sciences SCREEN-WELL Natural Product library (1 Library)
Supplier: Enzo Life Sciences BML28650100
Natural products are an unsurpassed source of chemical diversity and an ideal starting point for any screening program for pharmacologically active small molecules. Historically, natural products have been the most successful source of new drugs. The SCREEN-WELL Natural Product library offers over 500 isolated natural compounds of known structure and includes many compounds from ethnopharmacology. Compounds are provided in solution in DMSO. Individual compounds or subsets can be resupplied in gram quantities. Contents: 502 compounds including compounds from the following classes of products: Terpenoids, Peptolides, Flavones, Coumarins, Alkaloids, Macrolides, Isoflavones, Synthetic derivates and many more. Application: HTS. Use/Stability: Stable for at least one year from the date of receipt when stored at -80°C. Long Term Storage: -80°C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.