Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek
Ideal for peak identification work
Supplier: Restek Corporation 34466
Description
- CGA 165 connection and can deliver large volumes of sample
- Scotty™ (Air Liquide) 48
- Contents: 48L
- Pressure: 300 psig (21 bar)
- Outlet Fitting: CGA 165
- Weight: 1.75 lb. (0.8kg)
- Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
- DOT Specifications: 39 NRC
All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements
Specifications
DOT 39 NRC | |
10.2 cm | |
1000 ppm | |
Ideal for peak identification work | |
41 cm | |
1.75 lb. | |
0.8 kg |
Carbon Dioxide in Nitrogen | |
4 in. | |
48 L | |
16.25 in. | |
300 psi | |
12.68 gal. | |
Air Standard |
Safety and Handling
ShelfLife : 2 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction