Learn More
Scotsman™ Filter Cartridge
Keeps ice makers cleaner; reduce failures due to lime/scale, dirt and excessive chlorine
Supplier: Curran Taylor Inc RC20S/EMD
Description
- Complete with head and adaptors
- Includes scale inhibitor
Specifications
34,069 L | |
Ice makers | |
Built-in scale inhibitor, bacteriostatic Carbon block filtration medium | |
NSF, FDA CFR-21 |
9000 gal. | |
Reduces sediment and chlorine for equipment protection. Helps maintain efficiency with annual change. | |
Filter Cartridge for Model 121 | |
34069L (9000 gal.) |