Learn More
Atlantic Nuclear SCINTILLATION FLUID 4L CONTAIN
Supplier: Atlantic Nuclear 111175
Scintillation Fluid, Econo Safe Liquid Scintillation Fluid is formulated for use in Applications Requiring a General Purpose Cocktail, This Fluid can Accommodate Aqueous and Nonaqueous Samples, 4L