Learn More
Science First™ Simple Form Capacitor
Explore the principle of capacitance.
Supplier: Science First™ 6154730
Includes: Formed aluminum plates with binding posts (2), banana plug sockets and instructions
Description
- Simply charge the plates with a dielectric placed in between
- Used to show charge is contained on the dielectric and not the metal plates
Specifications
Simple Form Capacitor | |
2 Formed Aluminum plates with binding posts, banana plug sockets and instructions |