Learn More
Fine Science Tools Schwartz Micro Serrefines, Jaw: 10 x 1.75mm, 26 mm
Supplier: Fine Science Tools 1805202
Product: Schwartz Micro Serrefine - Light Bend/26mm/ Shape: Light Bend/ Jaw Dimensions: 10 x 1.75mm/ Clamping Pressure: Strong/ Alloy: Stainless Steel/ Length: 26mm/ Serrated: No/ Unit: Each