Learn More
Sartorius Vivaclear™ Centrifugal Filters
500 - 100μL For fast and reliable clarification / filtration.
Supplier: Sartorius VK01P042
Description
Vivaclear centrifugal filters are disposable microfiltration units for the fast and reliable clarification/filtration of biological samples in the range from 100 to 500μL. Vivaclear units feature a high-flux PES membrane of 0.8μm pore size and can be used in fixed angle rotors accepting 2.2 ml centrifuge tubes. Standard applications are sample clarification before loading onto Vivapure protein purification spin columns, particle removal, plasma or serum filtration, and removal of cells or cell debris from cell lysates or cell supernatants.
- High-flux, 0.8 mm PES membrane
- Used in fixed angle rotors
- Low hold-up volume
- Fast and reproducible performance
Specifications
Single-use | |
PES | |
0.8 μm | |
100/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.