Learn More
Sartorius arium comfort II Combined System (TYPE 1 - Ultrapure Water and TYPE 2 - EDI Water)
Use the Sartorius arium™ comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 2 pure water.
$19505.00
Specifications
Conductivity | 0.055 Product water, Less than1500 feed water |
---|---|
Bacterial Content | <1 CFU/L |
Daily Output | 120 (ASTM type 1), 10 (Type 2) |
Flow Rate | Up to 2 L/min. (ASTM type 1), Less than 3 L/min. (Type 2) |
Product Water Type | Type 1 and 2 |
Catalog Number | Mfr. No. | Mounting | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Mounting | Price | Quantity & Availability | |||||
14-556-202
|
Sartorius
H2OII2TOCB |
Wall |
Each for $19,505.00
|
|
|||||
14-556-204
|
Sartorius
H2OII2TOCT |
Benchtop |
Each for $19,505.00
|
|
|||||
14-556-203
|
Sartorius
H2OII2TOCD |
Underbench | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
arium comfort II systems produce ASTM Type 1 ultrapure water and Type 2 pure water. The systems features electrodeionization (EDI) technology, and a unique filter cartridge for producing ultrapure water of the highest quality, delivering consistently high-grade water quality at rates of up to 120l/h. The quality of this water even exceeds the ASTM Type 1 specifications. The system's integrated UV lamp (185/254 nm) prevents microbiological growth and reduces the TOC content to a minimum. The current TOC value is continuously checked by an integrated TOC monitor specially developed for this ultrapure water system and is continuously displayed.
- Flexible, space-saving twin technology
- Efficient iJust technology
- Uses disposable bagtank technology with dispense gun connections
- Built-in TOC monitor analyzes and displays actual TOC values
- Integrated UV lamp, 185/254nm, reduce the TOC ≤ 2ppb
- Convenient SD card slot
- User-friendly menu navigation
Benefits:
- Produces Type 2 and Type ASTM 1 water from tap water all in one unit; adapted to the customer's needs; the display will conveniently remain at eye level
- Optimized water consumption
- Meet customer's lab requirements and saves maintenance time
- Safe operation
- Reliable TOC reduction
- Supports reliable recording of important data
- Easy operation
- HPLC
- AAS
- Ion chromatography
- Photometry
- Microbiological media and reagents
- Histology
- ELISA, RIA
- Buffers and pH solutions
- Feed water for laboratory devices, such as autoclaves, glassware washers, humidifiers, water baths etc.
Specifications
0.055 Product water, Less than1500 feed water | |
120 (ASTM type 1), 10 (Type 2) | |
Type 1 and 2 | |
<2 product water, <1000 feed water | |
35.6°F to 86°F | |
Feed water for laboratory devices, such as autoclaves, Glassware washers, humidifiers, water baths etc | |
TOC monitor | |
4 to 10 | |
35.6°F to 95°F | |
<4 | |
17.12 in. | |
19.7 in. | |
18.74 in. | |
79.36 lb. | |
100/240 V 50/60 Hz | |
50/60 Hz |
<1 CFU/L | |
Up to 2 L/min. (ASTM type 1), Less than 3 L/min. (Type 2) | |
18.2 Mohm.cm product water | |
Arium Comfort II Combined System | |
2°C to 30°C | |
80% | |
<1/mL | |
5°C to 45°C | |
2°C to 35°C | |
<1 | |
43.5 cm | |
50.1 cm | |
47.6 cm | |
36 kg | |
100/240 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.