Learn More
Sartorius arium™ Comfort I Combined System (TYPE 1 - Ultrapure Water and TYPE 3 - RO Water)
Use the Sartorius arium™ Comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.
$12186.00 - $12780.00
Specifications
Conductivity | 0.055 Product water, Less than1500 feed water |
---|---|
Bacterial Content | <1 CFU/L |
Daily Output | 120 (ASTM type 1), 16 (Type 3) |
Product Water Type | Type 1 to 3 |
Resistivity | 18.2 Mohm.cm product water |
Catalog Number | Mfr. No. | Includes | Mounting | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Mounting | Price | Quantity & Availability | ||||
14-556-209
|
Sartorius
H2OI2TOCT |
TOC monitor | Benchtop |
Each for $12,780.00
|
|
||||
14-556-207
|
Sartorius
H2OI2TOCB |
TOC monitor | Wall |
Each for $12,780.00
|
|
||||
15-555-519
|
Sartorius
H2OI1TOCT |
UV lamp, TOC monitor | Benchtop |
Each for $12,186.00
|
|
||||
14-556-208
|
Sartorius
H2OI2TOCD |
Wall mounting kit for the display dispense unit and TOC monitor | Underbench | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
arium comfort I systems produce ASTM Type 1 ultrapure water and Type 3 pure water. The systems features the latest reverse osmosis technology, and a unique filter cartridge for producing ultrapure water of the highest quality, delivering consistently high-grade water quality at rates of up to 120l/h. The quality of this water even exceeds the ASTM Type 1 specifications. The system's integrated UV lamp (185/254 nm) prevents microbiological growth and reduces the TOC content to a minimum. The current TOC value is continuously checked by an integrated TOC monitor specially developed for this ultrapure water system and is continuously displayed.
- Reliable TOC content of <2 ppb for reproducible results
- Automatically optimised water consumption with iJust
- Easy-to-use - unique glass display with touch function
Benefits:
- Produces Type 3 and Type ASTM 1 water from tap water all in one unit; adapted to the customer's needs; the display will conveniently remain at eye level
- Optimized water consumption
- Meet customer's lab requirements and saves maintenance time
- Safe operation
- Reliable TOC reduction
- Supports reliable recording of important data
- Easy operation
- HPLC
- GC-MS, AAS, ICP-MS
- Ion chromatography
- TOC-Analysis
- Photometry
- Buffer and pH-solutions
- Feed water for ultrapure water systems, rinsing glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer
Specifications
0.055 Product water, Less than1500 feed water | |
120 (ASTM type 1), 16 (Type 3) | |
18.2 Mohm.cm product water | |
35.6°F to 86°F | |
80% | |
Adsorption by spherical activated Carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration | |
35.6°F to 95°F | |
<4 | |
17.12 in. | |
19.7 in. | |
18.74 in. | |
68.3 lb. | |
100/240 V 50/60 Hz |
<1 CFU/L | |
Type 1 to 3 | |
<2 product water, <2000 feed water | |
2°C to 30°C | |
4 to 10 | |
5°C to 45°C | |
2°C to 35°C | |
<1 | |
43.5 cm | |
50.1 cm | |
47.6 cm | |
31 kg |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.