Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix SARS-CoV-2 (Isolate: USA-WA1/2020) Culture Fluid (Heat Inactivated)
SARS-CoV-2 (Isolate: USA-WA1/2020) Culture Fluid is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus.
$257.00
Specifications
Content And Storage | Heat inactivated viral culture fluids should be stored at -65°C or below |
---|---|
Description | Repetitive freezing and thawing is not recommended (aliquot material if necessary) |
For Use With (Equipment) | To avoid cross-contamination, use separate pipette tips for all reagents |
Type | Culture Fluid |
For Use With (Application) | Isolate: USA-WA1/2020 |
Catalog Number | Mfr. No. | Quantity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Includes | Price | Quantity & Availability | ||||
22-158-364
|
ZeptoMetrix
0810587CFHI0.5ML |
0.5 mL | Each frozen aliquot contains 0.50 mL of heat inactivated viral culture fluid |
Each for $257.00
|
|
||||
22-158-369
|
ZeptoMetrix
0810587CFHI |
1 mL | Each frozen aliquot contains 1 mL of heat inactivated viral culture fluid |
Each for $469.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Heat inactivated viral culture fluids are sold as consumable testing materials
- The suitability and performance characteristics should be determined by your laboratory for each intended usage
- Viral inactivation is verified after heat inactivation by the absence of viral growth in tissue culture-based infectivity assays
Specifications
Heat inactivated viral culture fluids should be stored at -65°C or below | |
To avoid cross-contamination, use separate pipette tips for all reagents | |
Isolate: USA-WA1/2020 |
Repetitive freezing and thawing is not recommended (aliquot material if necessary) | |
Culture Fluid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title