Learn More
Bullard™ SALUS HC Powered Air Purifying Respirator System
SALUS HC high performing respiratory solution is designed to protect clinicians from exposure to particulates and biological aerosols including viruses and bacteria. Choose from two different sizes and systems that offer one or two HEPA filters and batteries.
Supplier: Bullard™ SALHC20LF2M
Description
- Designed for healthcare and EMS professionals
- New NIOSH PAPR 100-P standard
- Unique shoulder carriage design
- Compact, lightweight, comfortable
- Two speeds (low/high)
Includes:
- Medium or Large Narrow LF Hood
- Blower Motor
- Shoulder Carriage
- HEPA Filter - 1 or 2
- Battery - 1 or 2
- Battery Charger
Specifications
25 | |
Lithium-ion | |
PARTICULATE | |
PAPR Healthcare Systems and Kits | |
Loose fitting HEPA hood | |
Yes |
9.5 hours | |
7.5 to 15 in. | |
NIOSH PAPR100-P | |
Medium | |
Narrow LF Hood, Blower Motor, Shoulder Carriage, HEPA Filter, Battery, and Battery Charger | |
SALHC20LF2M |