Learn More
Cederlane SALMONELLA O SINGLE FACTOR 08
SDP
Supplier:  Cederlane 40224(SS)
Encompass
(Product: O:8),(Reacts with factor:C2, C3),(No. of tests: 150),(SSI� SALMONELLA O GROUP ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC0599909