Learn More
Cederlane SALMONELLA HMD
Supplier: Cederlane 40294(SS)
(Product: HMD),(Reacts with factor:1,2 1,5 1,6 1,7 z6),(No. of tests: 150),(SSI� SALMONELLA H PHASE POOL ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial