Learn More
Cederlane SALMONELLA 8MULTIPHASE HG
Supplier: Cederlane 40299(SS)
(Product: H:G),(Reacts with factor:All phases having g and m,t),(No. of tests: 150),(SSI� SALMONELLA H PHASE POOL ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial